Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Chloro-7-iodo-7-deazapurine | 123148-78-7 | 99.9% | C6H3ClIN3 | 10 G
SDP

Catalog No. 5000434997
Encompass_Preferred

6-Chloro-7-iodo-7-deazapurine is a biochemical reagent. It can be used as a biological material or organic compound for life science related research and serves as a pharmaceutical intermediate.

  • Biochemical reagent
  • Can be used as biological material for life science research
  • Can be used as organic compound for life science research
  • Used as pharmaceutical intermediate
  • Appearance: Solid, off-white to gray
  • Purity: 99.99%

Catalog No. 50-004-34997 Supplier Medchemexpress LLC Supplier No. HY7733610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Chloro-7-iodo-7-deazapurine is a biochemical reagent. It can be used as a biological material or organic compound for life science related research and serves as a pharmaceutical intermediate.

  • Biochemical reagent
  • Can be used as biological material for life science research
  • Can be used as organic compound for life science research
  • Used as pharmaceutical intermediate
  • Appearance: Solid, off-white to gray
  • Purity: 99.99%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.