Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. SPB05853DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SPB05853DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB05853DA Supplier Thermo Scientific Chemicals Supplier No. SPB05853DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 150020-64-7
Molecular Formula C5H2Cl2N2O2S2
Molecular Weight (g/mol) 257.103
InChI Key APRATEMYDZDYJD-UHFFFAOYSA-N
PubChem CID 2799717
IUPAC Name 6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride
SMILES C1=CSC2=NC(=C(N21)S(=O)(=O)Cl)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro-imidazo 2,1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro, acmc-1c5af, 6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonylchloride, 6-chloro-5-imidazo 2,1-b thiazolesulfonyl chloride, 6-chloro-imidazo 2, 1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride,6-chloro
Formula Weight 257.12
Percent Purity 97%
Chemical Name or Material 6-Chloroimidazo[2, 1-b][1, 3]thiazole-5-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.