Learn More
Accela Chembio Inc 6-chloroindole | 25g | 17422-33-2 | MFCD00005681 | 98% | Shelf Life: 1800 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50249786
Encompass_Preferred
6-chloroindole | 25g | 17422-33-2 | MFCD00005681 | 98% | Shelf Life: 1800 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50-249-786 Supplier Accela Chembio Inc Supplier No. SY00233825G
6-chloroindole | 25g | 17422-33-2 | MFCD00005681 | 98% | Shelf Life: 1800 Days | Light Sensitive/nitrogen Or Argon/+4