Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloroindole, 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
17422-33-2
C8H6ClN
151.6
MFCD00005681
YTYIMDRWPTUAHP-UHFFFAOYSA-N
6-chloroindole,1h-indole, 6-chloro,6-chloro indole,6-chloro-indole,6-chlor-1h-indole,pubchem1669,6-chloro-1-h-indole,#,ksc174m9p
87111
CHEBI:80918
6-chloro-1H-indole
C1=CC(=CC2=C1C=CN2)Cl
17422-33-2
C8H6ClN
151.6
MFCD00005681
YTYIMDRWPTUAHP-UHFFFAOYSA-N
6-chloroindole,1h-indole, 6-chloro,6-chloro indole,6-chloro-indole,6-chlor-1h-indole,pubchem1669,6-chloro-1-h-indole,#,ksc174m9p
87111
CHEBI:80918
6-chloro-1H-indole
C1=CC(=CC2=C1C=CN2)Cl
Quantity 250mg
Infrared Spectrum Authentic
Melting Point 88°C to 91°C
Color Beige
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Solubility Information Solubility in water: slightly soluble
Formula Weight 151.6
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 6-Chloroindole, 99%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 241-449-4