Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(chloromethyl)benzothiazole, 97%, Thermo Scientific™

Catalog No. AC441350010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441350010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC441350010 Supplier Thermo Scientific Chemicals Supplier No. 441350010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 149440-49-3
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.66
InChI Key WVJSICZOZHJCLJ-UHFFFAOYSA-N
Synonym 6-chloromethyl benzo d thiazole, 6-chloromethyl benzothiazole, benzothiazole, 6-chloromethyl, 6-chloromethyl-1,3-benzothiazole, acmc-20n5pd, 6-chloromethylbenzothiazole
PubChem CID 22905771
IUPAC Name 6-(chloromethyl)-1,3-benzothiazole
SMILES C1=CC2=C(C=C1CCl)SC=N2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 50°C to 52°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 183.66
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-(Chloromethyl)benzothiazole
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.