Learn More
Cayman Chemical 6-Chloropurine, Purity: ≥95%, CAS: 87-42-3, 5 g
SDP
Catalog No. 502918807
Encompass_Preferred
A synthetic intermediate; has been used in the synthesis of antibacterial or kinase-inhibiting purines, as well as anticancer agents; increases survival time in combination with azaserine in an S180 mouse model of sarcoma at 20 mg/kg per day
Catalog No. 50-291-8807 Supplier Cayman Chemical Supplier No. 388795 G
A synthetic intermediate; has been used in the synthesis of antibacterial or kinase-inhibiting purines, as well as anticancer agents; increases survival time in combination with azaserine in an S180 mouse model of sarcoma at 20 mg/kg per day