Learn More
Cayman Chemical 6-Chloropurine Riboside, Purity: ≥98%, CAS: 5399-87-1, 1 g
SDP
Catalog No. 502911097
Encompass_Preferred
A nucleoside precursor; used in the synthesis of adenosine or N6-benzyladenosine derivatives with anticancer or antiparasitic activities in vitro
Catalog No. 50-291-1097 Supplier Cayman Chemical Supplier No. 360021 G
A nucleoside precursor; used in the synthesis of adenosine or N6-benzyladenosine derivatives with anticancer or antiparasitic activities in vitro