Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Chloropyridin-3-amine | 5350-93-6 | 128.56 | 25 G
SDP

Catalog No. 5000449902
Encompass_Preferred

6-Chloropyridin-3-amine is a biochemical reagent used as a biological material or organic compound for various life science related research applications.

  • Appears as a solid, ranging from light brown to brown.
  • Purity verified by HPLC at 99.97%.
  • Features a melting point of 81-83°C.
  • Boiling point is 205.39°C at 760 mmHg.
  • Soluble in DMSO at 100 mg/mL.
  • 1H NMR spectrum is consistent with the chemical structure.

Catalog No. 50-004-49902 Supplier Medchemexpress LLC Supplier No. 25GHYW007481
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Chloropyridin-3-amine is a biochemical reagent used as a biological material or organic compound for various life science related research applications.

  • Appears as a solid, ranging from light brown to brown.
  • Purity verified by HPLC at 99.97%.
  • Features a melting point of 81-83°C.
  • Boiling point is 205.39°C at 760 mmHg.
  • Soluble in DMSO at 100 mg/mL.
  • 1H NMR spectrum is consistent with the chemical structure.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.