Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloropyridine-2-carboxylic acid, 97%

Catalog No. AC436660050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436660050 5 g Glass bottle
AC436660010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC436660050 Supplier Thermo Scientific Chemicals Supplier No. 436660050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 4684-94-0 | C6H4ClNO2 | 157.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4684-94-0
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.56
InChI Key ZLKMOIHCHCMSFW-UHFFFAOYSA-N
Synonym 6-chloropicolinic acid, 2-chloro-6-pyridinecarboxylic acid, 6-chloro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-chloro, 2-chloropyridine-6-carboxylic acid, 6-chloro-2-picolinic acid, 6-cpa, picolinic acid, 6-chloro, 6-chloro-2-carboxy-pyridine, caswell no. 214b
PubChem CID 20812
IUPAC Name 6-chloropyridine-2-carboxylic acid
SMILES C1=CC(=NC(=C1)Cl)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 162.6°C to 162.7°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 157.56
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloropyridine-2-carboxylic acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.