Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Chloropyrimidine-4-carboxylic acid | 37131-91-2 | 158.54 | 250 MG
SDP

Catalog No. 5000450078
Encompass_Preferred

6-Chloropyrimidine-4-carboxylic acid is a drug intermediate for synthesis of various active compounds.

  • Molecular formula: C5H3ClN2O2
  • Target: Drug intermediate
  • Pathway: Others
  • Storage at -20°C, sealed, away from moisture
  • In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
  • Appearance: Off-white to yellow solid
  • 1H NMR spectrum consistent with structure
  • Water (KF): 0.15%
  • Complies with given specifications

Catalog No. 50-004-50078 Supplier Medchemexpress LLC Supplier No. 250MGHYW005983
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Chloropyrimidine-4-carboxylic acid is a drug intermediate for synthesis of various active compounds.

  • Molecular formula: C5H3ClN2O2
  • Target: Drug intermediate
  • Pathway: Others
  • Storage at -20°C, sealed, away from moisture
  • In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
  • Appearance: Off-white to yellow solid
  • 1H NMR spectrum consistent with structure
  • Water (KF): 0.15%
  • Complies with given specifications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.