Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Chloropyrimidine-4-carboxylic acid | 37131-91-2 | 158.54 | 500 MG
SDP

Catalog No. 5000448588
Encompass_Preferred

6-Chloropyrimidine-4-carboxylic acid is a drug intermediate used for the synthesis of various active compounds. It is supplied as an off-white to yellow solid with a purity of 98.72%.

  • Drug intermediate for synthesis of various active compounds
  • Off-white to yellow solid appearance
  • Purity of 98.72%
  • Molecular weight of 158.54

Catalog No. 50-004-48588 Supplier Medchemexpress LLC Supplier No. HYW005983500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Chloropyrimidine-4-carboxylic acid is a drug intermediate used for the synthesis of various active compounds. It is supplied as an off-white to yellow solid with a purity of 98.72%.

  • Drug intermediate for synthesis of various active compounds
  • Off-white to yellow solid appearance
  • Purity of 98.72%
  • Molecular weight of 158.54

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.