Learn More
Medchemexpress LLC 6-Chloropyrimidine-4-carboxylic acid | 37131-91-2 | MFCD08692928 | 158.54 | 5 G

Supplier: Medchemexpress LLC HYW0059835G
6-Chloropyrimidine-4-carboxylic acid is an off-white to yellow solid with a molecular weight of 158.54 and a molecular formula of C₅H₃ClN₂O₂. This chemical is ideal for research applications, offering high purity and specific storage requirements to maintain its integrity.
- Off-white to yellow solid appearance
- High-performance liquid chromatography (HPLC) purity of 98.72%
- Molecular weight of 158.54
- Store at -20°C, sealed and away from moisture
- For solutions, store at -80°C for up to 6 months or -20°C for up to 1 month, sealed and away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.