Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloroquinoline 98.0+%, TCI America™
SDP

Catalog No. C02835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C02835G 5 g
C02831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C02835G Supplier TCI America Supplier No. C02835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 612-57-7
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.60
MDL Number MFCD00024024
InChI Key GKJSZXGYFJBYRQ-UHFFFAOYSA-N
Synonym quinoline, 6-chloro, unii-k4a4bau5se, ccris 3981, k4a4bau5se, 6-chloro-quinoline, 6-chlor-chinolin;, zlchem 764, pubchem5813, acmc-1aulp, quinoline, 6-chloro-,
PubChem CID 69163
IUPAC Name 6-chloroquinoline
SMILES ClC1=CC=C2N=CC=CC2=C1

Specifications

Melting Point 41°C
Boiling Point 264°C
Quantity 5 g
Formula Weight 163.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 6-Chloroquinoline
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.