Learn More
Cayman Chemical 6-ChrysenamIn 10mg
SDP
Catalog No. 502911212
Encompass_Preferred
A carcinogen and anticancer agent; neonatal administration induces the formation of hepatomas and pulmonary adenocarcinomas in adult mice at 600 UG/animal; s.c.; inhibits tumor growth in an EO771 murine mammary carcinoma model from 3-110 mg/kg
Catalog No. 50-291-1212 Supplier Cayman Chemical Supplier No. 3578010 MG
A carcinogen and anticancer agent; neonatal administration induces the formation of hepatomas and pulmonary adenocarcinomas in adult mice at 600 UG/animal; s.c.; inhibits tumor growth in an EO771 murine mammary carcinoma model from 3-110 mg/kg