Learn More
AdipoGen 6-Cl-2-OCH3-9-phenoxyacridine
SDP
Catalog No. 502561440
Encompass_Preferred
Chemical. CAS 7478-26-4. Formula C20H14ClNO2. MW 335.78. Synthetic. Acridine. Halogenated heterocycle used as building block.
Catalog No. 50-256-1440 Supplier AdipoGen Supplier No. CDXC0178G001
Chemical. CAS 7478-26-4. Formula C20H14ClNO2. MW 335.78. Synthetic. Acridine. Halogenated heterocycle used as building block.