Learn More
Chemglass Life Sciences Clip Plates for 10-13mm Tubes, Each Holds 6 Tubes, 6/Pkg (Fig. 1)
SDP
Catalog No. 501214944
Encompass
Combines rocking and rotating in one compact and versatile unit.Easily converts to rocking to rotating (or vice versa) in just 5 seconds.Magnetic stainless steel platform and clip plate System holds almost any container at any angle and expansion kits available for increased capacity and different containers.Magnetic Strips hold bags for complete and even soaking of membrane-based blot assays/washings.Ideal for Cold Rooms and Incubators and Maintains set speed between 0-38 degreesC.Optional 3-D orbital shaker attachment.Supplied with 1 clip plate each for 10-13 mm, 15-17mm, 28-30mm tubes, 4 bag mounting strips and a non-skid tray.
Catalog No. 50-121-4944 Supplier Chemglass Life Sciences Supplier No. CLS3960005
Combines rocking and rotating in one compact and versatile unit.Easily converts to rocking to rotating (or vice versa) in just 5 seconds.Magnetic stainless steel platform and clip plate System holds almost any container at any angle and expansion kits available for increased capacity and different containers.Magnetic Strips hold bags for complete and even soaking of membrane-based blot assays/washings.Ideal for Cold Rooms and Incubators and Maintains set speed between 0-38 degreesC.Optional 3-D orbital shaker attachment.Supplied with 1 clip plate each for 10-13 mm, 15-17mm, 28-30mm tubes, 4 bag mounting strips and a non-skid tray.