Learn More
Waters Corp Waters Rebuild Kit, 6 Position Select Valve, HPLC System
Catalog No. 50792384 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit is designed for the 2790/2795 Six Column Select Valve. It facilitates the necessary maintenance by allowing for the replacement of the rotor seal and stator face assembly.
  • Rebuild kit for maintenance of select valves.
  • Compatible with HPLC systems.
  • Replacement is recommended as required.
  • Includes one unit per package.
Catalog No. 50-792-384 Supplier Waters Corp Supplier No. 700000434
This rebuild kit is designed for the 2790/2795 Six Column Select Valve. It facilitates the necessary maintenance by allowing for the replacement of the rotor seal and stator face assembly.
  • Rebuild kit for maintenance of select valves.
  • Compatible with HPLC systems.
  • Replacement is recommended as required.
  • Includes one unit per package.