Learn More
Waters Corp Waters Column Select Valve, 6-Position, 14-Port
Catalog No. 50792385 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 6-Column Selection Valve is designed for precise control in chromatographic systems, allowing users to select one of six columns at a time. This valve enhances experimental flexibility by providing an option for bypass or waste in one of its positions.
  • Enables selection of one of six columns at a time.
  • One position configurable for bypass or waste.
  • Requires two additional column clips for installation.
  • Includes slots in the front of the column heater to hold tubing in place.
Catalog No. 50-792-385 Supplier Waters Corp Supplier No. 700000435
Add to Cart
Add to Cart
Request a Quote
The 6-Column Selection Valve is designed for precise control in chromatographic systems, allowing users to select one of six columns at a time. This valve enhances experimental flexibility by providing an option for bypass or waste in one of its positions.
  • Enables selection of one of six columns at a time.
  • One position configurable for bypass or waste.
  • Requires two additional column clips for installation.
  • Includes slots in the front of the column heater to hold tubing in place.