Learn More
Felix Storch Inc ACCUCOLD 6 CU FT MOMCUBE BREAST MILK
SDP
Catalog No. NC3070024
Encompass_Preferred
The MLRS62BIADAMC is sized at just under 32" hight to fit under lower counters complying with ADA guidelines for accessibility. It is designed for freestanding or recessed installation. It comes in a white cabinet finish with a blue accented control panel. An LED display that allows the user to monitor and change the temperature as needed. Several alarms ensure temperature stability, and the factory-installed external door lock offers added security. The self-closing, reversible door allows for flexibility in unit placement. The factory installed antimicrobial handle is powder-coated with naturally occurring silver ions, which help to reduce the spread of germs, allowing for a safer, cleaner user experience, while the hospital grade cord with 'green dot' plug is also included for added safety: https://www.accucold.com/product/MLRS62BIADAMC
Catalog No. NC3070024 Supplier Felix Storch Inc Supplier No. MLRS62BIADAMC
The MLRS62BIADAMC is sized at just under 32" hight to fit under lower counters complying with ADA guidelines for accessibility. It is designed for freestanding or recessed installation. It comes in a white cabinet finish with a blue accented control panel. An LED display that allows the user to monitor and change the temperature as needed. Several alarms ensure temperature stability, and the factory-installed external door lock offers added security. The self-closing, reversible door allows for flexibility in unit placement. The factory installed antimicrobial handle is powder-coated with naturally occurring silver ions, which help to reduce the spread of germs, allowing for a safer, cleaner user experience, while the hospital grade cord with 'green dot' plug is also included for added safety: https://www.accucold.com/product/MLRS62BIADAMC