Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[6]Cycloparaphenylene 98.0+%, TCI America™
SDP

Catalog No. C338620MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C338620MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C338620MG Supplier TCI America Supplier No. C338620MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 156980-13-1
Molecular Formula C36H24
Synonym [6]CPP

Specifications

Quantity 20 mg
Formula Weight 456.59
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [6]Cycloparaphenylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.