Learn More
Cayman Chemical 6-DeoxypencIclovIr 10mg
SDP
Catalog No. 502909908
Encompass_Preferred
An inactive metabolite of famciclovir; a precursor metabolite to penciclovir; has been used in the synthesis of prodrug forms of penciclovir
Catalog No. 50-290-9908 Supplier Cayman Chemical Supplier No. 3560210 MG
An inactive metabolite of famciclovir; a precursor metabolite to penciclovir; has been used in the synthesis of prodrug forms of penciclovir