Learn More
Cayman Chemical 6-Deoxypenciclovir, Purity: ≥95%, CAS: 104227-86-3, 5 mg
SDP
Catalog No. 502909954
Encompass_Preferred
An inactive metabolite of famciclovir; a precursor metabolite to penciclovir; has been used in the synthesis of prodrug forms of penciclovir
Catalog No. 50-290-9954 Supplier Cayman Chemical Supplier No. 356025 MG
An inactive metabolite of famciclovir; a precursor metabolite to penciclovir; has been used in the synthesis of prodrug forms of penciclovir