Learn More
Selleck Chemical LLC 6-Diazo-5-oxo-L-norleucine
SDP
Catalog No. 5000002039
Encompass_Preferred
6-Diazo-5-oxo-L-norleucine (Diazooxonorleucine L-6-Diazo-5-oxonorleucine DON) an antibiotic isolated from Streptomyces is a glutaminase antagonist with IC50 of 1 mM for cKGA (kidney-type glutaminase) 6-Diazo-5-oxo-L-norleucine exhibits analgesic antibacterial antiviral and anticancer properties
Catalog No. 50-000-02039 Supplier Selleck Chemical LLC Supplier No. S862025MG
6-Diazo-5-oxo-L-norleucine (Diazooxonorleucine L-6-Diazo-5-oxonorleucine DON) an antibiotic isolated from Streptomyces is a glutaminase antagonist with IC50 of 1 mM for cKGA (kidney-type glutaminase) 6-Diazo-5-oxo-L-norleucine exhibits analgesic antibacterial antiviral and anticancer properties