Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-(difluoromethoxy)-2-[(3,4-dimethoxy-2-pyridinyl)methylsulfanyl]-1H-benzimidazole | 102625-64-9 | MFCD07368273 | 98.8% | 367.37 | C16H15F2N3O3S | 10 G
SDP

Catalog No. 5000299937
Encompass_Preferred

Pantoprazole sulfide is the sulfide metabolite of the proton-pump inhibitor pantoprazole, supplied as a research-grade compound for use as a reference standard, impurity profiling, and metabolic studies. The compound has formula C16H15F2N3O3S and molecular weight 367.37, and is provided with a certificate of analysis and safety documentation.

  • Purity 98.83%
  • White to off-white powder physical form
  • Suitable as a reference standard and for impurity and metabolite studies
  • Available in multiple pack sizes including 10 g
  • Storage: powder at -20°C (3 years) or 4°C (2 years)
  • Documentation includes COA, SDS, and data sheet

Catalog No. 50-002-99937 Supplier Medchemexpress LLC Supplier No. HYW01603310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pantoprazole sulfide is the sulfide metabolite of the proton-pump inhibitor pantoprazole, supplied as a research-grade compound for use as a reference standard, impurity profiling, and metabolic studies. The compound has formula C16H15F2N3O3S and molecular weight 367.37, and is provided with a certificate of analysis and safety documentation.

  • Purity 98.83%
  • White to off-white powder physical form
  • Suitable as a reference standard and for impurity and metabolite studies
  • Available in multiple pack sizes including 10 g
  • Storage: powder at -20°C (3 years) or 4°C (2 years)
  • Documentation includes COA, SDS, and data sheet

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.