Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-(difluoromethoxy)-2-[(3,4-dimethoxy-2-pyridinyl)methylsulfanyl]-1H-benzimidazole | 102625-64-9 | MFCD07368273 | 98.8% | 367.37 g/mol | C16H15F2N3O3S | 50 G
SDP

Catalog No. 5000299712
Encompass_Preferred

Pantoprazole sulfide is the desulfide metabolite of the proton-pump inhibitor pantoprazole, supplied as a research-grade chemical for analytical, metabolic, and impurity studies. It is intended for laboratory research use and not for human or clinical application.

  • Metabolite of pantoprazole.
  • Molecular formula C16H15F2N3O3S; molecular weight 367.37 g/mol.
  • High purity (98.8%) suitable for analytical applications.
  • Supplied as a powder for stable handling and storage.
  • Storage recommendations: powder -20°C (long term), 4°C (short term); in solvent -80°C (6 months), -20°C (1 month).
  • Intended for research use only.

Catalog No. 50-002-99712 Supplier Medchemexpress LLC Supplier No. HYW01603350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pantoprazole sulfide is the desulfide metabolite of the proton-pump inhibitor pantoprazole, supplied as a research-grade chemical for analytical, metabolic, and impurity studies. It is intended for laboratory research use and not for human or clinical application.

  • Metabolite of pantoprazole.
  • Molecular formula C16H15F2N3O3S; molecular weight 367.37 g/mol.
  • High purity (98.8%) suitable for analytical applications.
  • Supplied as a powder for stable handling and storage.
  • Storage recommendations: powder -20°C (long term), 4°C (short term); in solvent -80°C (6 months), -20°C (1 month).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.