Learn More
Bioworld 6-Dimethylallylamino Purine Riboside, 100 mg
Supplier:  Bioworld 404500041
Encompass_Preferred
6-Dimethylallylamino purine ribosideSynonyms: 2iP Riboside, 6-(3,3-Dimethylallylamino)purine riboside, N6-(γ,γ-Dimethylallyl)adenosine, N6-(2-Isopentenyl)adenosineCytokin growth regulatorAppearance: white powderMelting Point: 143-146 °CSoluble in waterAppearance: Solid
Catalog No. 50-255-887