Learn More
Apexbio Technology LLC 6-(Dimethylamino)purine 938-55-6 500mg
SDP
Catalog No. 5000093327
Encompass_Preferred
6-(Dimethylamino)purine (CAS 938-55-6) is a small molecule inhibitor targeting serine/threonine protein kinases modulating cell cycle progression and gene expression Studies have shown that it suppresses prolactin-induced expression of milk protein-related genes in mammary epithelial cells and disrupts meiotic maturation in mammalian oocytes Additionally it induces apoptosis in lymphoma cells by downregulating the expression of key cell cycle-associated genes including proliferating cell nuclear antigen insulin-induced gene 1 and serine protease inhibitor 2 This compound is utilized in research focused on cell cycle regulation and apoptosis
Catalog No. 50-000-93327 Supplier Apexbio Technology LLC Supplier No. C6989500
6-(Dimethylamino)purine (CAS 938-55-6) is a small molecule inhibitor targeting serine/threonine protein kinases modulating cell cycle progression and gene expression Studies have shown that it suppresses prolactin-induced expression of milk protein-related genes in mammary epithelial cells and disrupts meiotic maturation in mammalian oocytes Additionally it induces apoptosis in lymphoma cells by downregulating the expression of key cell cycle-associated genes including proliferating cell nuclear antigen insulin-induced gene 1 and serine protease inhibitor 2 This compound is utilized in research focused on cell cycle regulation and apoptosis