Learn More
Medchemexpress LLC 6-(Dimethylamino)purine | 938-55-6 | 99.9% | 163.18 | 1 ML
SDP
Catalog No. 5000453585
Encompass_Preferred
6-(Dimethylamino)purine is a serine threonine protein kinase inhibitor that can affect various cellular processes. It is used in research to study cellular mechanisms.
  • Inhibits prolactin-induced expression of lactoprotein genes in rabbit mammary gland cells.
  • Affects the maturation of mammalian oocytes.
  • Leads to downregulation of genes related to cell proliferation and cell cycle progression.
  • Induces apoptosis in lymphoma cells.
Catalog No. 50-004-53585 Supplier Medchemexpress LLC Supplier No. HYW01012810MM/1ML
6-(Dimethylamino)purine is a serine threonine protein kinase inhibitor that can affect various cellular processes. It is used in research to study cellular mechanisms.
  • Inhibits prolactin-induced expression of lactoprotein genes in rabbit mammary gland cells.
  • Affects the maturation of mammalian oocytes.
  • Leads to downregulation of genes related to cell proliferation and cell cycle progression.
  • Induces apoptosis in lymphoma cells.