Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(Dimethylamino)purine 98.0+%, TCI America™
SDP

Catalog No. D38941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38941G 1 g
D38945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38941G Supplier TCI America Supplier No. D38941G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 938-55-6
Molecular Formula C7H9N5
Molecular Weight (g/mol) 163.184
MDL Number MFCD00005573
InChI Key BVIAOQMSVZHOJM-UHFFFAOYSA-N
PubChem CID 3134
ChEBI CHEBI:60281
IUPAC Name N,N-dimethyl-7H-purin-6-amine
SMILES CN(C)C1=NC=NC2=C1NC=N2

Specifications

Melting Point 260°C
Color White-Yellow
Quantity 1 g
Synonym 6-dimethylaminopurine, 6-dimethylamino purine, n,n-dimethyladenine, n6,n6-dimethyladenine, 6-dimethyladenine, dimethyladenine, n 6 ,n 6-dimethyladenine, n,n-dimethyl-9h-purin-6-amine, adenine, n,n-dimethyl, 6-dmap
Formula Weight 163.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-(Dimethylamino)purine
RTECSNumber UO7440636
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.