Learn More
Cayman Chemical 6-(Dimethylamino)purine, Purity: ≥98%, CAS: 938-55-6, 100 mg
SDP
Catalog No. 502916038
Encompass_Preferred
A non-selective protein kinase inhibitor; enhances the inactivation of MAPK and induces the formation of pronuclei in primary mouse oocytes; induces pronuclear formation, cortical contraction, and cortical granule breakdown in Xenopus oocytes
Catalog No. 50-291-6038 Supplier Cayman Chemical Supplier No. 38963100 MG
A non-selective protein kinase inhibitor; enhances the inactivation of MAPK and induces the formation of pronuclei in primary mouse oocytes; induces pronuclear formation, cortical contraction, and cortical granule breakdown in Xenopus oocytes