Learn More
AdipoGen 6-Dimethylaminopurine
SDP
Catalog No. 502561122
Encompass_Preferred
Chemical. CAS 938-55-6. Formula C7H9N5. MW 163.18. Synthetic. A puromycin analog that acts as a protein kinase inhibitor. Selectively induces premature mitosis in mammalian cells. Inhibits TNF-alpha upregulation of ICAM-1, lipolysis, and the activation of JNK/SAPK. Inhibits p34cdc2/cyclin B IC50 = 300 µM. Induces apoptosis in S-phase-arrested HeLa cells ~5 mM.
Catalog No. 50-256-1122 Supplier AdipoGen Supplier No. CDXD0318G001
Chemical. CAS 938-55-6. Formula C7H9N5. MW 163.18. Synthetic. A puromycin analog that acts as a protein kinase inhibitor. Selectively induces premature mitosis in mammalian cells. Inhibits TNF-alpha upregulation of ICAM-1, lipolysis, and the activation of JNK/SAPK. Inhibits p34cdc2/cyclin B IC50 = 300 µM. Induces apoptosis in S-phase-arrested HeLa cells ~5 mM.