Learn More
Selleck Chemical LLC 6-Dimethylaminopurine
SDP
Catalog No. 5000002908
Encompass_Preferred
6-Dimethylaminopurine (N N-Dimethyladenine) is a serine threonine protein kinase inhibitor It inhibits the germinal vesicle breakdown and the meiotic maturation of oocytes
Catalog No. 50-000-02908 Supplier Selleck Chemical LLC Supplier No. S608825MG
6-Dimethylaminopurine (N N-Dimethyladenine) is a serine threonine protein kinase inhibitor It inhibits the germinal vesicle breakdown and the meiotic maturation of oocytes