Learn More
AdipoGen 6-ECDCA
SDP
Catalog No. 502562289
Encompass_Preferred
Chemical. CAS 459789-99-2. Formula C26H44O4. MW 420.6. Potent and selective FXR agonist EC50= 99nM. Induces SHP in HSCs to suppress TIMP-1 expression. Apoptosis inducer. Protects against liver fibrosis development in rat in vivo. Displays anticholeretic activity in rat in vivo. Promotes preadipocyte differentiation. Regulates adipogenesis and insulin signaling in vivo. Inhibits vascular smooth muscle cell inflammation and migration.
Catalog No. 50-256-2289 Supplier AdipoGen Supplier No. AGCR13560M100
Chemical. CAS 459789-99-2. Formula C26H44O4. MW 420.6. Potent and selective FXR agonist EC50= 99nM. Induces SHP in HSCs to suppress TIMP-1 expression. Apoptosis inducer. Protects against liver fibrosis development in rat in vivo. Displays anticholeretic activity in rat in vivo. Promotes preadipocyte differentiation. Regulates adipogenesis and insulin signaling in vivo. Inhibits vascular smooth muscle cell inflammation and migration.