Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Ethoxy-2-methylquinolin-4-ol

Catalog No. OTV0008741G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0008741G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0008741G Supplier Sigma Aldrich Supplier No. OTV0008741G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-penten-2-one is volatile compound present in berry fruit of two aronia melanocarpa genotypes
  • Urinary 3-penten-2-one is a useful biomarker for increased accumulation of acetaldehyde during abnormal metabolic stress.3-penten-2-one was used in the synthesis of the alkaloids (+/-)-senepodine g and (+/-)-cermizine c.3-penten-2-one inhibits nitric oxide production and inducible nitric oxide synthase expression via heme oxygenase-1 expression in lipopolysaccharide-activated raw264.7 macrophages.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • 145017.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

CAS 15644-91-4
Molecular Formula C12H13NO2
Linear Formula C12H13NO2
MDL Number MFCD02368819
Quantity 5 g
Molecular Weight (g/mol) 203.24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.