Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Ethoxy-2-naphthaleneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. E11075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1107-5G 5 g
E11071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1107-5G Supplier TCI America Supplier No. E11075G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 352525-98-5
Molecular Formula C12H13BO3
Molecular Weight (g/mol) 216.04
MDL Number MFCD03427009
InChI Key INXXVGFSXYJGHI-UHFFFAOYSA-N
IUPAC Name (6-ethoxynaphthalen-2-yl)boronic acid
SMILES CCOC1=CC2=CC=C(C=C2C=C1)B(O)O

Specifications

Melting Point 255°C
Color White
Quantity 5 g
Synonym 6-ethoxy-2-naphthaleneboronic acid, 6-ethoxynaphthalen-2-yl boronic acid, 6-ethoxynaphthalene-2-boronic acid, boronic acid, 6-ethoxy-2-naphthalenyl, 2-ethoxy-6-naphthaleneboronic acid
Formula Weight 216.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.