Learn More
Sigma Aldrich 6-Ethyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Reaction of fecl2 or fecl3 with sodium ethanethiolate in n-methylformamide (nmf) has been evaluated
- Reaction of sodium ethanethiolate with substituted 1,2-dithiole-3-thiones has been studied.sodium ethanethiolate may be used in the equilibrium titrations in n-methylformamide of g-25 gel filtered (ox)-state femo cofactor [femoco(ox)] from azotobacter vinelandii nitrogenase
- It may be used in the synthesis of a series of hybrid compounds combining the pharmacophoric redox moieties of vitamin e and key features responsible for the antiarrhythmic properties of the class i antiarrhythmics procainamide and lidocaine
Specifications
Specifications
| CAS | 53939-83-6 |
| Molecular Formula | C6H8N2OS |
| Linear Formula | C6H8N2OS |
| MDL Number | MFCD02186898 |
| Quantity | 10 g |
| Molecular Weight (g/mol) | 156.21 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.