Learn More
Medchemexpress LLC 6α-ethyl-3α,7α,23-trihydroxy-24-nor-5β-cholan-23-sulfate | 1000403-03-1 | >98.0% | 494.66 g·mol⁻1 | C25H43NaO6S | 50MG
SDP
Catalog No. 5000235405
Encompass_Preferred
INT-767 is a synthetic dual agonist of the farnesoid X receptor (FXR) and the G protein-coupled bile acid receptor TGR5 (GPBAR1), used as a research reagent in preclinical studies of liver and metabolic disease. Reported activity includes mean EC50s of 30 nM (FXR) and 630 nM (TGR5); CAS 1000403-03-1; molecular formula C25H43NaO6S; molecular weight 494.66 g·mol⁻1. Typical supplier-reported purity is >98%. Material is for research use only and not for human therapeutic use.
  • Dual FXR and TGR5 agonist for bile-acid signaling studies.
  • Well-characterized potency: EC50 30 nM (FXR) and 630 nM (TGR5).
  • High reported purity suitable for in vitro and in vivo experiments.
  • Supplied as the sodium-sulfate derivative with defined molecular weight.
  • Widely used in preclinical models of liver injury, steatosis, and metabolic regulation.
  • For research use only; not for clinical or human use.
Catalog No. 50-002-35405 Supplier Medchemexpress LLC Supplier No. HY1243450MG
INT-767 is a synthetic dual agonist of the farnesoid X receptor (FXR) and the G protein-coupled bile acid receptor TGR5 (GPBAR1), used as a research reagent in preclinical studies of liver and metabolic disease. Reported activity includes mean EC50s of 30 nM (FXR) and 630 nM (TGR5); CAS 1000403-03-1; molecular formula C25H43NaO6S; molecular weight 494.66 g·mol⁻1. Typical supplier-reported purity is >98%. Material is for research use only and not for human therapeutic use.
  • Dual FXR and TGR5 agonist for bile-acid signaling studies.
  • Well-characterized potency: EC50 30 nM (FXR) and 630 nM (TGR5).
  • High reported purity suitable for in vitro and in vivo experiments.
  • Supplied as the sodium-sulfate derivative with defined molecular weight.
  • Widely used in preclinical models of liver injury, steatosis, and metabolic regulation.
  • For research use only; not for clinical or human use.