Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Ethyl-4-hydroxy-2H-chromen-2-one

Catalog No. JRD11081G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD11081G 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD11081G Supplier Sigma Aldrich Supplier No. JRD11081G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-(diethoxymethyl)imidazole is an orthoamide protected imidazole
  • The dianion reacts selectively at the 2 position 1-(diethoxymethyl)imidazole is a 1-substituted imidazoles
  • Isoprene-mediated lithiation of 1-(diethoxymethyl)imidazole, leads to the preparation of the corresponding 2-lithio intermediate, which reacts with different electrophiles to afford 2-functionalised imidazoles.1-(diethoxymethyl)imidazole may be used in the preparation of various hydroxy- and amino-functionalized imidazoles, via isoprene-mediated lithiation followed by reaction with an electrophile

Specifications

CAS 5505-28-2
Molecular Formula C11H10O3
Linear Formula C11H10O3
MDL Number MFCD03424528
Quantity 10 mL
Molecular Weight (g/mol) 190.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.