Learn More
AdipoGen 6-FAM
SDP
Supplier:  AdipoGen CDXC0016M050
Encompass_Preferred
Chemical. CAS 3301-79-9. Formula C21H12O7. MW 376.32. Synthetic. The amine-reactive fluorescein derivatives are probably the most common fluorescent derivatization reagents for covalently labeling proteins. Carboxyfluorescein is better retained in cells than is fluorescein, its pKa of ~6.5 is lower than the cytosolic pH of most cells pH ~6.8-7.4. Carboxyfluorescein is commonly employed as a polar tracer.
Catalog No. 50-256-1546