Learn More
Abcam 6-FAM (6-Carbo x yfluorescein), Amine-reactive fluorescent label, 200MG
Catalog No. 502705316
Encompass_Preferred
MW 376.3 Da, Purity >97%. Amine-reactive fluorescent label. 5-FAM (ab145293) isomer. 6-CFDA (ab145298) and 6-CFDA N-succinimidyl ester (ab145296) hydrolytic product. Polar tracer. Covalently labels peptides and proteins. Deprotonates below pH 7 resulting in reduced fluorescence. Compatible with most fluorescence detection equipment.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5316 Supplier Abcam Supplier No. AB145294200MG
MW 376.3 Da, Purity >97%. Amine-reactive fluorescent label. 5-FAM (ab145293) isomer. 6-CFDA (ab145298) and 6-CFDA N-succinimidyl ester (ab145296) hydrolytic product. Polar tracer. Covalently labels peptides and proteins. Deprotonates below pH 7 resulting in reduced fluorescence. Compatible with most fluorescence detection equipment.
The product is subject to the following: Abcam Restricted Use Statement