Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 6-FAM amidite | 200 mg | CAS 204697-37-0 | MDL MFCD01940925

Catalog No. 502355557
Encompass

6-FAM amidite is a Amidite reagent (Subcategory: Dye Amidite) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 204697-37-0
- MDL: MFCD01940925
- InChIKey: MGPYJVWEJNTXLC-UHFFFAOYSA-N
- Molecular Weight: 843.954761998
- Molecular Formula: C46H58N3O10P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29322090
- Country Of Origin: China
- IUPAC: 6-((6-(((2-cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)hexyl)carbamoyl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl bis(2,2-dimethylpropanoate)
- SMILES: CC(C)N(C(C)C)P(OCCCCCCNC(C1=CC2=C(C(OC23C4=C(OC5=C3C=CC(OC(C(C)(C)C)=O)=C5)C=C(OC(C(C)(C)C)=O)C=C4)=O)C=C1)=O)OCCC#N

Catalog No. 50-235-5557 Supplier STA Pharmaceutical Supplier No. STAAM10105200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-FAM amidite is a Amidite reagent (Subcategory: Dye Amidite) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 204697-37-0
- MDL: MFCD01940925
- InChIKey: MGPYJVWEJNTXLC-UHFFFAOYSA-N
- Molecular Weight: 843.954761998
- Molecular Formula: C46H58N3O10P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29322090
- Country Of Origin: China
- IUPAC: 6-((6-(((2-cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)hexyl)carbamoyl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl bis(2,2-dimethylpropanoate)
- SMILES: CC(C)N(C(C)C)P(OCCCCCCNC(C1=CC2=C(C(OC23C4=C(OC5=C3C=CC(OC(C(C)(C)C)=O)=C5)C=C(OC(C(C)(C)C)=O)C=C4)=O)C=C1)=O)OCCC#N

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.