Learn More
Medchemexpress LLC 6-FAM-PEG3-azide | 412319-45-0 | 98.1% | C29H28N4O9 | 10MG
SDP
Catalog No. 5000259993
Encompass_Preferred
6-FAM-PEG3-Azide is a fluorescent labeling reagent composed of a 6-carboxyfluorescein (6-FAM) dye linked to an azide via a PEG3 spacer. It is designed for covalent conjugation to alkyne-functionalized partners using copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC), enabling fluorescent labeling of oligonucleotides and other biomolecules.
  • Fluorescent labeling with excitation 498 nm and emission 532 nm.
  • PEG3 spacer improves water solubility and reduces steric hindrance.
  • Azide functional group for CuAAC and SPAAC click chemistry.
  • Solid, light yellow to yellow form stable when stored as powder at -20°C.
  • High purity (~98.1%) suitable for analytical and labeling applications.
Catalog No. 50-002-59993 Supplier Medchemexpress LLC Supplier No. HY13583510MG
6-FAM-PEG3-Azide is a fluorescent labeling reagent composed of a 6-carboxyfluorescein (6-FAM) dye linked to an azide via a PEG3 spacer. It is designed for covalent conjugation to alkyne-functionalized partners using copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC), enabling fluorescent labeling of oligonucleotides and other biomolecules.
  • Fluorescent labeling with excitation 498 nm and emission 532 nm.
  • PEG3 spacer improves water solubility and reduces steric hindrance.
  • Azide functional group for CuAAC and SPAAC click chemistry.
  • Solid, light yellow to yellow form stable when stored as powder at -20°C.
  • High purity (~98.1%) suitable for analytical and labeling applications.