Learn More
Apexbio Technology LLC 6 FAM SE, 50mg. Cas: 92557-81-8 MFCD: MFCD00077323. Used for labeling DNA, protein and peptide.
SDP
Supplier:  Apexbio Technology LLC A877150
Encompass_Preferred
6-FAM SE, also known as 6-Carboxyfluorescein N-hydroxysuccinimide ester or 6-Carboxyfluorescein N-succinimidyl ester, is an isomer of carboxyfluorescein and amine-reactive succinimidyl ester carboxyfluorescein. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5189