Learn More
Electron Microscopy Sciences Curved Fine Forceps with Non-serrated Tips, 152 mm OAL
SDP
Catalog No. NC0918819
Encompass_Preferred
EMS Brand, Swiss made forceps from low carbon austenitic steel, corrosion resistance, high quality, curved tips, available with different length and tips style
OAL: 152 mm
Catalog No. NC0918819 Supplier Electron Microscopy Sciences Supplier No. 729116
EMS Brand, Swiss made forceps from low carbon austenitic steel, corrosion resistance, high quality, curved tips, available with different length and tips style
OAL: 152 mm