Learn More
AdipoGen 6-FITC
SDP
Supplier:  AdipoGen CDXF0111M250
Encompass_Preferred
Chemical. CAS 18861-78-4. Formula C21H11NO5S. MW 389.39. Synthetic. 6-Fluorescein isothiocyanate 6-FITC is an amine-reactive derivative of fluorescein dye that has wide-ranging applications as a label for antibodies and other probes, for use in fluorescence microscopy, flow cytometry and immunofluorescence-based assays such as Western blotting and ELISA. Spectral data lambdaex=492nm, lambdaem=518nm.
Catalog No. 50-256-0870