Learn More
Ace Glass, Inc. Column extender, 6" ID x 18" , 6" O-ring grooved flat flange top, 6" ground flat flange bottom
SDP
Catalog No. 50972403
Encompass_Preferred
6 I.D. x 18 column extender with size 6 o-ring grooved flat flange top and size 6 ground flat flange bottom. Column extender can be mated to column with ACE 6517 series clamps.
Catalog No. 50-972-403 Supplier Ace Glass, Inc. Supplier No. 586278
6 I.D. x 18 column extender with size 6 o-ring grooved flat flange top and size 6 ground flat flange bottom. Column extender can be mated to column with ACE 6517 series clamps.