Learn More
AdipoGen 6-(Fluorescein-5(6)-carboxamido) hexanoic acid (50 mg)
SDP
Catalog No. 501490381
Encompass_Preferred
6-(Fluorescein-5(6)-carboxamido) hexanoic acid (50 mg), AdipoGen Life Sciences. Chemical. CAS: 265981-56-4. Formula: C27H23NO8. MW: 489.47. Synthetic. Used as a fluorescein-based fluorescent label for proteins that can be analyzed in applications such as quenching experiments. Spectral data: lambdaex 491nm; lambdaem 515nm (0.1 M Tris, pH 9.0).
Catalog No. 50-149-0381 Supplier AdipoGen Supplier No. CDXF0015M050
6-(Fluorescein-5(6)-carboxamido) hexanoic acid (50 mg), AdipoGen Life Sciences. Chemical. CAS: 265981-56-4. Formula: C27H23NO8. MW: 489.47. Synthetic. Used as a fluorescein-based fluorescent label for proteins that can be analyzed in applications such as quenching experiments. Spectral data: lambdaex 491nm; lambdaem 515nm (0.1 M Tris, pH 9.0).