Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Fluoro-1,2,3,4-tetrahydroquinoline, 97%

Catalog No. p-7032899
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH35364MD 250 mg
AAH3536403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH35364MD Supplier Thermo Scientific Chemicals Supplier No. H35364MD
Add to Cart

CAS: 59611-52-8 | C9H10FN | 151.18 g/mol

6-Fluoro-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoro-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59611-52-8
Molecular Formula C9H10FN
Molecular Weight (g/mol) 151.18
MDL Number MFCD09040997
InChI Key NECDDBBJVCNJNS-UHFFFAOYSA-N
Synonym quinoline, 6-fluoro-1,2,3,4-tetrahydro, 6-fluoro-1,2,3,4-tetrahydro-quinoline
PubChem CID 14783911
IUPAC Name 6-fluoro-1,2,3,4-tetrahydroquinoline
SMILES FC1=CC2=C(NCCC2)C=C1

Specifications

Melting Point 32°C to 34°C
Quantity 250 mg
Formula Weight 151.18
Percent Purity 97%
Chemical Name or Material 6-Fluoro-1,2,3,4-tetrahydroquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.