Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Fluoro-2-methyl-4-quinolinol 98.0+%, TCI America™
SDP

Catalog No. F09971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F09971G Supplier TCI America Supplier No. F09971G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15912-68-2
Molecular Formula C10H8FNO
Molecular Weight (g/mol) 177.178
MDL Number MFCD00041442
InChI Key BKXCHVFCJZJATJ-UHFFFAOYSA-N
Synonym 6-fluoro-2-methylquinolin-4-ol, 6-fluoro-2-methylquinolin-4 1h-one, 6-fluoro-4-hydroxy-2-methylquinoline, 6-fluoro-2-methyl-4-quinolinol, 4-quinolinol, 6-fluoro-2-methyl, 6-fluoro-2-methyl-quinolin-4-ol, acmc-1aerw, maybridge1_000260, acmc-209j2u, 6-fluoro-4-hydroxy quinilidine
PubChem CID 85184
IUPAC Name 6-fluoro-2-methyl-1H-quinolin-4-one
SMILES CC1=CC(=O)C2=C(N1)C=CC(=C2)F

Specifications

Melting Point 266°C
Color White
Quantity 1 g
Formula Weight 177.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluoro-2-methyl-4-quinolinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.