Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Fluoro-3-pyridinylboronic acid, 97%, Thermo Scientific™

Catalog No. AC397160050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397160050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC397160050 Supplier Thermo Scientific Chemicals Supplier No. 397160050

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 351019-18-6
Molecular Formula C5H5BFNO2
Molecular Weight (g/mol) 140.91
MDL Number MFCD03411559
InChI Key OJBYZWHAPXIJID-UHFFFAOYSA-N
Synonym 2-fluoropyridine-5-boronic acid, 2-fluoro-5-pyridylboronic acid, 6-fluoropyridin-3-yl boronic acid, 6-fluoropyridine-3-boronic acid, 2-fluoro-5-pyridineboronic acid, 2-fluoropyridin-5-ylboronic acid, 2-fluoropyridin-5-boronic acid, 6-fluoro-3-pyridinylboronic acid, 6-fluoropyridin-3-yl-3-boronic acid, 6-fluoro-3-pyridineboronic acid
PubChem CID 2783397
IUPAC Name (6-fluoropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC=C(F)N=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 172.0°C to 176.0°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 140.91
Percent Purity 97%
Chemical Name or Material 6-Fluoro-3-pyridinylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.